Product Details

SNP ID
rs7930940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60758313 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGTTCCTTTATAACAGTTAAACA[A/G]GGTGCCACAGAAACATACATTAAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MS4A15 PubMed Links

Gene Details

Gene
MS4A15
Gene Name
membrane spanning 4-domains A15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098835.1 Intron NP_001092305.1
NM_001278242.1 Intron NP_001265171.1
NM_152717.2 Intron NP_689930.1
XM_006718459.3 Intron XP_006718522.1
XM_011544812.2 Intron XP_011543114.1
XM_011544814.1 Intron XP_011543116.1

View Full Product Details