Product Details

SNP ID
rs7949780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65114136 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAGTGCCAGGTGCCCACCCTGG[A/G]TCTGGGTCCTGGCCTGATGACAGGC
Phenotype
MIM: 134690 MIM: 606866 MIM: 603414 MIM: 615738 MIM: 604575
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAU PubMed Links

Gene Details

Gene
FAU
Gene Name
FAU, ubiquitin like and ribosomal protein S30 fusion
There are no transcripts associated with this gene.

Gene
MRPL49
Gene Name
mitochondrial ribosomal protein L49
There are no transcripts associated with this gene.

Gene
TM7SF2
Gene Name
transmembrane 7 superfamily member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277233.1 Intron NP_001264162.1
NM_003273.3 Intron NP_003264.2
Gene
VPS51
Gene Name
VPS51, GARP complex subunit
There are no transcripts associated with this gene.

Gene
ZNHIT2
Gene Name
zinc finger HIT-type containing 2
There are no transcripts associated with this gene.

View Full Product Details