Product Details

SNP ID
rs8117656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:32828388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTGTAAATATATTCCAAAGTCT[A/G]TTTCACTTAGTTAATTGGAAAATGT
Phenotype
MIM: 603108
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAPRE1 PubMed Links

Gene Details

Gene
MAPRE1
Gene Name
microtubule associated protein RP/EB family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012325.2 Intron NP_036457.1
XM_011528696.2 Intron XP_011526998.1

View Full Product Details