Product Details

SNP ID
rs7798757
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92114371 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGATATCCAAAGCAATCTTGGAAA[A/T]AGAACTGAGTTGGAGGACATAAAAC
Phenotype
MIM: 604001 MIM: 601637
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AKAP9 PubMed Links

Gene Details

Gene
AKAP9
Gene Name
A-kinase anchoring protein 9
There are no transcripts associated with this gene.

Gene
CYP51A1
Gene Name
cytochrome P450 family 51 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000786.3 Intron NP_000777.1
NM_001146152.1 Intron NP_001139624.1

View Full Product Details