Product Details

SNP ID
rs8077269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:33530046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAAAGTAGGTCCAGGAAATACAT[A/G]AATAAATGCGAAGGCCATAAAGCGT
Phenotype
MIM: 601784
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AA06 PubMed Links

Gene Details

Gene
AA06
Gene Name
uncharacterized LOC100506677
There are no transcripts associated with this gene.

Gene
ASIC2
Gene Name
acid sensing ion channel subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001094.4 Intron NP_001085.2
NM_183377.1 Intron NP_899233.1

View Full Product Details