Product Details

SNP ID
rs8074165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:31392747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCGGTGCCGCCAGTAGGGTGGAC[A/G]GAGTCATATCACTAAACGCACACTA
Phenotype
MIM: 611999
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB11FIP4 PubMed Links

Gene Details

Gene
RAB11FIP4
Gene Name
RAB11 family interacting protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303542.2 Intron NP_001290471.2
NM_032932.5 Intron NP_116321.2

View Full Product Details