Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001193314.1 | 2009 | UTR 3 | NP_001180243.1 | ||
NM_001193315.1 | 2009 | UTR 3 | NP_001180244.1 | ||
NM_001193316.1 | 2009 | UTR 3 | NP_001180245.1 | ||
NM_001193317.1 | 2009 | UTR 3 | NP_001180246.1 | ||
NM_022067.3 | 2009 | UTR 3 | NP_071350.2 | ||
XM_011537066.1 | 2009 | UTR 3 | XP_011535368.1 | ||
XM_017021579.1 | 2009 | UTR 3 | XP_016877068.1 | ||
XM_017021580.1 | 2009 | Intron | XP_016877069.1 | ||
XM_017021581.1 | 2009 | Intron | XP_016877070.1 |