Product Details

SNP ID
rs7074880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:1042625 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTTGTATGGTTGTGTAGGAGAGC[C/T]GGTTACCTCTTCCAAGGGAATTCCT
Phenotype
MIM: 604055 MIM: 615391
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IDI1 PubMed Links

Gene Details

Gene
IDI1
Gene Name
isopentenyl-diphosphate delta isomerase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317955.1 Intron NP_001304884.1
NM_001317956.1 Intron NP_001304885.1
NM_001317957.1 Intron NP_001304886.1
NM_004508.3 Intron NP_004499.2
XM_011519457.1 Intron XP_011517759.1
XM_017016191.1 Intron XP_016871680.1
Gene
IDI2-AS1
Gene Name
IDI2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details