Product Details

SNP ID
rs6943934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:1149542 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGACAGACACCATGAAATAAGCA[C/T]AGCAACGGTCACATGTCACCCACCG
Phenotype
MIM: 610699
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFAND2A PubMed Links

Gene Details

Gene
ZFAND2A
Gene Name
zinc finger AN1-type containing 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182491.2 3538 Intron NP_872297.2
XM_017012774.1 3538 UTR 3 XP_016868263.1

View Full Product Details