Product Details

SNP ID
rs6477989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113166820 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATGAGAGACCCTCTCTTTTCACA[A/G]CTTGAGCTCCTGGCCTTTCTCCTCT
Phenotype
MIM: 603088
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FKBP15 PubMed Links

Gene Details

Gene
FKBP15
Gene Name
FK506 binding protein 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015258.1 Intron NP_056073.1
XM_006717018.2 Intron XP_006717081.1
XM_006717019.1 Intron XP_006717082.1
XM_017014552.1 Intron XP_016870041.1
Gene
SLC31A2
Gene Name
solute carrier family 31 member 2
There are no transcripts associated with this gene.

View Full Product Details