Product Details

SNP ID
rs7857409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133036382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACTTGATGAGGCGTGCTCACAAC[A/T]AAATCTCCCCCAAAGGTCAGCTTTC
Phenotype
MIM: 604890
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GTF3C5 PubMed Links

Gene Details

Gene
GTF3C5
Gene Name
general transcription factor IIIC subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122823.1 Intron NP_001116295.1
NM_001286709.1 Intron NP_001273638.1
NM_012087.3 Intron NP_036219.2
XM_005272234.3 Intron XP_005272291.1
XM_011519201.1 Intron XP_011517503.1
XM_017015315.1 Intron XP_016870804.1
XM_017015316.1 Intron XP_016870805.1
Gene
LOC105376306
Gene Name
uncharacterized LOC105376306
There are no transcripts associated with this gene.

View Full Product Details