Product Details
- SNP ID
-
rs8070024
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:70080290 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATACCAAGATCTCTGAGAAACCATG[A/T]GATTAACATCGTATACCTTTTAGAA
- Phenotype
-
MIM: 605722
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KCNJ16
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116603573] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KCNJ16
- Gene Name
- potassium voltage-gated channel subfamily J member 16
View Full Product Details