Product Details

SNP ID
rs7211359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:64146394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTGGCGCCGGGGGTCAATCTGCA[A/G]CGCTAGAGCATGTGCTTGCGCATAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNHG25 PubMed Links

Gene Details

Gene
SNHG25
Gene Name
small nucleolar RNA host gene 25
There are no transcripts associated with this gene.

Gene
SNORA50C
Gene Name
small nucleolar RNA, H/ACA box 50C
There are no transcripts associated with this gene.

Gene
SNORD104
Gene Name
small nucleolar RNA, C/D box 104
There are no transcripts associated with this gene.

Gene
TEX2
Gene Name
testis expressed 2
There are no transcripts associated with this gene.

View Full Product Details