Product Details

SNP ID
rs6453465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:79615370 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGAAAAACAGCATATTTTTTATA[A/G]GTATAAACAAAAGTATCTAGGCTTT
Phenotype
MIM: 614121
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAPD4 PubMed Links

Gene Details

Gene
PAPD4
Gene Name
PAP associated domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114393.1 Intron NP_001107865.1
NM_001114394.1 Intron NP_001107866.1
NM_001297744.1 Intron NP_001284673.1
NM_001297745.1 Intron NP_001284674.1
NM_173797.3 Intron NP_776158.2
XM_011543219.2 Intron XP_011541521.1
XM_011543220.2 Intron XP_011541522.1
XM_011543221.2 Intron XP_011541523.1
XM_011543222.2 Intron XP_011541524.1
XM_011543223.2 Intron XP_011541525.1
XM_011543224.2 Intron XP_011541526.1
XM_011543225.2 Intron XP_011541527.1
XM_011543226.2 Intron XP_011541528.1
XM_011543227.2 Intron XP_011541529.1
XM_011543228.2 Intron XP_011541530.1
XM_011543229.2 Intron XP_011541531.1
XM_011543230.2 Intron XP_011541532.1
XM_011543231.2 Intron XP_011541533.1
XM_011543232.2 Intron XP_011541534.1
XM_011543233.2 Intron XP_011541535.1
XM_017009151.1 Intron XP_016864640.1
XM_017009152.1 Intron XP_016864641.1
XM_017009153.1 Intron XP_016864642.1
XM_017009154.1 Intron XP_016864643.1
XM_017009155.1 Intron XP_016864644.1
XM_017009156.1 Intron XP_016864645.1
XM_017009157.1 Intron XP_016864646.1
XM_017009158.1 Intron XP_016864647.1
XM_017009159.1 Intron XP_016864648.1
XM_017009160.1 Intron XP_016864649.1
XM_017009161.1 Intron XP_016864650.1
XM_017009162.1 Intron XP_016864651.1

View Full Product Details