Product Details
- SNP ID
-
hCV29407588
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:111644192 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACAGTCTCAGGGATGTCTGTTGCT[C/T]TGAAGGTCACTTGCCCCTCTTGCTG
- Phenotype
-
MIM: 604986
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BRAP
PubMed Links
Gene Details
- Gene
- BRAP
- Gene Name
- BRCA1 associated protein
There are no transcripts associated with this gene.
View Full Product Details