Product Details

SNP ID
rs7592580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27898323 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGACCTAGCCTATCTTGCATTTTA[C/T]TGCTAGGAACAAACACATTAAGGAT
Phenotype
MIM: 610497 MIM: 611132
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRE PubMed Links

Gene Details

Gene
BRE
Gene Name
brain and reproductive organ-expressed (TNFRSF1A modulator)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261840.1 Intron NP_001248769.1
NM_004899.4 Intron NP_004890.2
NM_199191.2 Intron NP_954661.1
NM_199192.2 Intron NP_954662.1
NM_199193.2 Intron NP_954663.1
NM_199194.2 Intron NP_954664.1
Gene
BRE-AS1
Gene Name
BRE antisense RNA 1
There are no transcripts associated with this gene.

Gene
RBKS
Gene Name
ribokinase
There are no transcripts associated with this gene.

View Full Product Details