Product Details

SNP ID
rs4711165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28147406 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACAATATCAAGTGCTAGTCAGAT[G/T]GGAAAACAGTTTTGCAGTTTCTTAT
Phenotype
MIM: 602240
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZKSCAN8 PubMed Links

Gene Details

Gene
ZKSCAN8
Gene Name
zinc finger with KRAB and SCAN domains 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278119.1 Intron NP_001265048.1
NM_001278121.1 Intron NP_001265050.1
NM_001278122.1 Intron NP_001265051.1
NM_006298.3 Intron NP_006289.2
XM_011514870.2 Intron XP_011513172.1
XM_011514873.2 Intron XP_011513175.1
XM_011514874.2 Intron XP_011513176.1
XM_017011266.1 Intron XP_016866755.1

View Full Product Details