Product Details

SNP ID
rs6902679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43174535 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGGCTGGTTGACTGGGCCGGGGAC[A/T]ACCTTACAAGCCAGCCGCTGCCCTT
Phenotype
MIM: 607489 MIM: 600589
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CUL9 PubMed Links

Gene Details

Gene
CUL9
Gene Name
cullin 9
There are no transcripts associated with this gene.

Gene
SRF
Gene Name
serum response factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292001.1 Intron NP_001278930.1
NM_003131.3 Intron NP_003122.1

View Full Product Details