Product Details

SNP ID
rs6853522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:153785068 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCCTGTCTTCAGGGGAGTGACTTG[A/C]AACTTATGGAACATATCACATGTGT
Phenotype
MIM: 604157
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SFRP2 PubMed Links

Gene Details

Gene
SFRP2
Gene Name
secreted frizzled related protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003013.2 Intron NP_003004.1

View Full Product Details