Product Details

SNP ID
rs7998003
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:20584971 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCTACAATGCATAGGGCAGCCCC[C/T]GGTAATAAAGAGCTATCCAGCCGAA
Phenotype
MIM: 600595
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFT88 PubMed Links

Gene Details

Gene
IFT88
Gene Name
intraflagellar transport 88
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318491.1 Intron NP_001305420.1
NM_001318493.1 Intron NP_001305422.1
NM_006531.4 Intron NP_006522.2
NM_175605.4 Intron NP_783195.2
XM_005266553.2 Intron XP_005266610.1
XM_006719870.3 Intron XP_006719933.1
XM_011535241.2 Intron XP_011533543.1
XM_011535242.1 Intron XP_011533544.1
XM_011535243.1 Intron XP_011533545.1
XM_017020757.1 Intron XP_016876246.1
XM_017020758.1 Intron XP_016876247.1
XM_017020759.1 Intron XP_016876248.1
XM_017020760.1 Intron XP_016876249.1
XM_017020761.1 Intron XP_016876250.1
XM_017020762.1 Intron XP_016876251.1
XM_017020763.1 Intron XP_016876252.1
XM_017020764.1 Intron XP_016876253.1
XM_017020765.1 Intron XP_016876254.1
XM_017020766.1 Intron XP_016876255.1
XM_017020767.1 Intron XP_016876256.1
XM_017020768.1 Intron XP_016876257.1
XM_017020769.1 Intron XP_016876258.1
XM_017020770.1 Intron XP_016876259.1
XM_017020771.1 Intron XP_016876260.1
XM_017020772.1 Intron XP_016876261.1
XM_017020773.1 Intron XP_016876262.1
XM_017020774.1 Intron XP_016876263.1
XM_017020775.1 Intron XP_016876264.1
XM_017020776.1 Intron XP_016876265.1

View Full Product Details