Product Details
- SNP ID
-
rs7985552
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:20706599 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGGGAGGAAATAATGAATTTAGTA[A/T]CAAGCCTGTTGAGTTTTGCCTGCCT
- Phenotype
-
MIM: 607587
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IL17D
PubMed Links
Gene Details
- Gene
- IL17D
- Gene Name
- interleukin 17D
View Full Product Details