Product Details

SNP ID
rs6925115
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:151454701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTTTGTTAATTTCTCAACAGAA[A/G]CATAATAAATAGTTCTTGAAGGTAT
Phenotype
MIM: 616332 MIM: 614917
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ARMT1 PubMed Links

Gene Details

Gene
ARMT1
Gene Name
acidic residue methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286562.1 Intron NP_001273491.1
NM_024573.2 Intron NP_078849.1
Gene
RMND1
Gene Name
required for meiotic nuclear division 1 homolog
There are no transcripts associated with this gene.

View Full Product Details