Product Details

SNP ID
rs6935618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:143453062 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGAATTAGTGAAGGGGAACTGGA[A/G]TTCAAACCCAGGTGTGTTGGACCAC
Phenotype
MIM: 615388 MIM: 603164
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAT2 PubMed Links

Gene Details

Gene
ADAT2
Gene Name
adenosine deaminase, tRNA specific 2
There are no transcripts associated with this gene.

Gene
PEX3
Gene Name
peroxisomal biogenesis factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003630.2 Intron NP_003621.1

View Full Product Details