Product Details

SNP ID
rs6568718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:112108824 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGGGAAAAAGTAGTTGATTTCCT[G/T]TGTTTCTTATTTAGAAAAAATTAAA
Phenotype
MIM: 600133
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM229B PubMed Links

Gene Details

Gene
FAM229B
Gene Name
family with sequence similarity 229 member B
There are no transcripts associated with this gene.

Gene
LAMA4
Gene Name
laminin subunit alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105206.2 6479 UTR 3 NP_001098676.2
NM_001105207.2 6479 UTR 3 NP_001098677.2
NM_001105208.2 6479 Intron NP_001098678.1
NM_001105209.2 6479 Intron NP_001098679.1
NM_002290.4 6479 UTR 3 NP_002281.3
XM_005266983.3 6479 UTR 3 XP_005267040.2
XM_005266984.3 6479 UTR 3 XP_005267041.2
XM_017010854.1 6479 UTR 3 XP_016866343.1

View Full Product Details