Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001260474.1 | 1895 | UTR 3 | NP_001247403.1 | ||
NM_001260475.1 | 1895 | UTR 3 | NP_001247404.1 | ||
NM_001260476.1 | 1895 | UTR 3 | NP_001247405.1 | ||
NM_001260477.1 | 1895 | UTR 3 | NP_001247406.1 | ||
NM_018669.5 | 1895 | UTR 3 | NP_061139.2 | ||
NM_033661.4 | 1895 | Intron | NP_387510.1 | ||
XM_011529433.1 | 1895 | UTR 3 | XP_011527735.1 | ||
XM_017028262.1 | 1895 | UTR 3 | XP_016883751.1 | ||
XM_017028263.1 | 1895 | UTR 3 | XP_016883752.1 | ||
XM_017028264.1 | 1895 | Intron | XP_016883753.1 |