Product Details

SNP ID
rs7508949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:20052723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGCTCTGTCGAGCCGTGGCGCC[C/G]TGCAAGGGAGTAAGAACGGAGCTCC
Phenotype
MIM: 610952
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CFAP61 PubMed Links

Gene Details

Gene
CFAP61
Gene Name
cilia and flagella associated protein 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167816.1 135 Intron NP_001161288.1
NM_015585.3 135 Intron NP_056400.3
XM_005260688.3 135 Intron XP_005260745.1
XM_005260689.2 135 Intron XP_005260746.1
XM_005260690.3 135 Intron XP_005260747.1
XM_005260691.3 135 Intron XP_005260748.1
XM_005260692.3 135 Intron XP_005260749.1
XM_005260693.3 135 Intron XP_005260750.1
XM_005260695.4 135 Intron XP_005260752.1
XM_005260696.4 135 Intron XP_005260753.1
XM_005260697.1 135 Intron XP_005260754.1
XM_011529211.1 135 Intron XP_011527513.1
XM_011529212.2 135 Intron XP_011527514.1
XM_011529213.1 135 Intron XP_011527515.1
XM_017027786.1 135 Intron XP_016883275.1
XM_017027787.1 135 Intron XP_016883276.1
XM_017027788.1 135 Intron XP_016883277.1
XM_017027789.1 135 Intron XP_016883278.1
XM_017027790.1 135 Intron XP_016883279.1
XM_017027791.1 135 Intron XP_016883280.1
XM_017027792.1 135 Intron XP_016883281.1
XM_017027793.1 135 Intron XP_016883282.1
XM_017027794.1 135 Intron XP_016883283.1
XM_017027795.1 135 Intron XP_016883284.1
Gene
CRNKL1
Gene Name
crooked neck pre-mRNA splicing factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278625.1 135 Intron NP_001265554.1
NM_001278626.1 135 Intron NP_001265555.1
NM_001278627.1 135 Intron NP_001265556.1
NM_001278628.1 135 Intron NP_001265557.1
NM_016652.5 135 Missense Mutation CGG,GGG R35G NP_057736.4

View Full Product Details