Product Details

SNP ID
rs7353635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:10639505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGTGCCCGCGGTCTGCTATACG[A/G]TGTACTCCATTCGGTTTAAGCTCTG
Phenotype
MIM: 601920
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
JAG1 PubMed Links

Gene Details

Gene
JAG1
Gene Name
jagged 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000214.2 4166 Missense Mutation ACC,ATC T1217I NP_000205.1
XM_017027707.1 4166 Missense Mutation ACC,ATC T1058I XP_016883196.1

View Full Product Details