Product Details

SNP ID
rs6114817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:2537179 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCCCAGTTTGAGGCAGCCACAAA[C/T]CAGCTGAGGGGTCCCACAGTGACCG
Phenotype
MIM: 606707
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMC2 PubMed Links

Gene Details

Gene
TMC2
Gene Name
transmembrane channel like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080751.2 Intron NP_542789.2
XM_005260660.3 Intron XP_005260717.1

View Full Product Details