Product Details
- SNP ID
-
rs6721427
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:144679434 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACAAATTTATAAACACTATAGGTAA[A/G]TATTAGATTGAAGCAAATAGATGAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TEX41
PubMed Links
Gene Details
- Gene
- TEX41
- Gene Name
- testis expressed 41 (non-protein coding)
There are no transcripts associated with this gene.
View Full Product Details