Product Details

SNP ID
rs7358020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:135286562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACATTGTGGTAGTGGGTACTCA[A/G]GAGAGAAGCAACAAGAATGGGACAA
Phenotype
MIM: 314997
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF75D PubMed Links

Gene Details

Gene
ZNF75D
Gene Name
zinc finger protein 75D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185063.2 3947 Intron NP_001171992.1
NM_007131.4 3947 UTR 3 NP_009062.2
XM_005262469.1 3947 UTR 3 XP_005262526.1
XM_005262471.3 3947 UTR 3 XP_005262528.1
XM_011531391.1 3947 UTR 3 XP_011529693.1
XM_011531395.2 3947 UTR 3 XP_011529697.1
XM_011531396.2 3947 UTR 3 XP_011529698.1
XM_011531397.2 3947 UTR 3 XP_011529699.1
XM_017029818.1 3947 UTR 3 XP_016885307.1
XM_017029819.1 3947 UTR 3 XP_016885308.1
XM_017029820.1 3947 UTR 3 XP_016885309.1

View Full Product Details