Product Details

SNP ID
rs9914988
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28856086 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATATGAGAAAACTAAAGGGCTCAG[A/G]GAGTTACTTTATAAATTAATGTAAA
Phenotype
MIM: 607435 MIM: 612070 MIM: 612071
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERAL1 PubMed Links

Gene Details

Gene
ERAL1
Gene Name
Era like 12S mitochondrial rRNA chaperone 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317985.1 Intron NP_001304914.1
NM_001317986.1 Intron NP_001304915.1
NM_005702.3 Intron NP_005693.1
Gene
FAM222B
Gene Name
family with sequence similarity 222 member B
There are no transcripts associated with this gene.

Gene
MIR144
Gene Name
microRNA 144
There are no transcripts associated with this gene.

Gene
MIR451A
Gene Name
microRNA 451a
There are no transcripts associated with this gene.

Gene
MIR451B
Gene Name
microRNA 451b
There are no transcripts associated with this gene.

Gene
MIR4732
Gene Name
microRNA 4732
There are no transcripts associated with this gene.

View Full Product Details