Product Details
- SNP ID
-
rs9297205
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:33549882 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGCCCAATACATTTGGTGATGTGG[C/T]ACTTAATACCAACTAGAGGTAGGAT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RNF122
PubMed Links
Gene Details
- Gene
- RNF122
- Gene Name
- ring finger protein 122
View Full Product Details