Product Details

SNP ID
rs10420848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57154520 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGATAGAGGAAAGAATGTAAGAG[A/G]TCAGCTTATATTCACAAACATGGAC
Phenotype
MIM: 611168
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUXA PubMed Links

Gene Details

Gene
DUXA
Gene Name
double homeobox A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012729.1 Intron NP_001012747.1
Gene
ZIM3
Gene Name
zinc finger imprinted 3
There are no transcripts associated with this gene.

View Full Product Details