Product Details

SNP ID
rs9653626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45416920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCATATCCCTGGAGAGGCAAATA[C/G]CAAACAATTAATGCTTATTTGGAGG
Phenotype
MIM: 611453 MIM: 610272
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DBNDD2 PubMed Links

Gene Details

Gene
DBNDD2
Gene Name
dysbindin domain containing 2
There are no transcripts associated with this gene.

Gene
MIR6812
Gene Name
microRNA 6812
There are no transcripts associated with this gene.

Gene
PIGT
Gene Name
phosphatidylinositol glycan anchor biosynthesis class T
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184728.2 Intron NP_001171657.1
NM_001184729.2 Intron NP_001171658.1
NM_001184730.2 Intron NP_001171659.1
NM_015937.5 Intron NP_057021.2
XM_005260430.2 Intron XP_005260487.1
XM_005260432.2 Intron XP_005260489.1
Gene
SYS1-DBNDD2
Gene Name
SYS1-DBNDD2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details