Product Details

SNP ID
rs6632767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:16130280 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGAGTACAATAACTACACCTTGGT[A/T]GATGTTTGAGCAGTGACAAAAGGAT
Phenotype
MIM: 305670
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GRPR PubMed Links

Gene Details

Gene
GRPR
Gene Name
gastrin releasing peptide receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005314.2 Intron NP_005305.1

View Full Product Details