Product Details

SNP ID
rs7548422
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:145994000 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGCTTAGGACAAATTTAACAGTAA[C/A]GATGACAATCCTCACCTCAGTATAA
Phenotype
MIM: 606599
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
TXNIP PubMed Links

Gene Details

Gene
TXNIP
Gene Name
thioredoxin interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001313972.1 1470 Intron NP_001300901.1
NM_006472.5 1470 Intron NP_006463.3
XM_017000085.1 1470 Missense Mutation GTT,TTT V386F XP_016855574.1

View Full Product Details