Product Details

SNP ID
rs9456113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:168444463 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGTAATGATCTCTCTTCTTTCCA[C/T]CAAAGTCATTCTTTAGAAATGGCAC
Phenotype
MIM: 607223
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMOC2 PubMed Links

Gene Details

Gene
SMOC2
Gene Name
SPARC related modular calcium binding 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166412.1 Intron NP_001159884.1
NM_022138.2 Intron NP_071421.1
XM_011536065.1 Intron XP_011534367.1
XM_011536066.1 Intron XP_011534368.1

View Full Product Details