Product Details

SNP ID
rs10417943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55013969 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCCTTCAGAAAGTAAATATGAGA[A/G]GGGTGGAGACGGTGCATTATCTTAT
Phenotype
MIM: 605546
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GP6 PubMed Links

Gene Details

Gene
GP6
Gene Name
glycoprotein VI platelet
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083899.2 2000 UTR 3 NP_001077368.2
NM_001256017.2 2000 UTR 3 NP_001242946.2
NM_016363.5 2000 UTR 3 NP_057447.5

View Full Product Details