Product Details
- SNP ID
-
rs6122392
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:63196115 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTTCAGTGGGCAACAGATCTGGAA[A/G]GAAAGATTTTCAAAAAAAAAAAAAA
- Phenotype
-
MIM: 616529
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
YTHDF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs200141223] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- YTHDF1
- Gene Name
- YTH N6-methyladenosine RNA binding protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017798.3 |
2567 |
UTR 3 |
|
|
NP_060268.2 |
View Full Product Details