Product Details

SNP ID
rs5975745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:136536886 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAATATGCAAATTTGTGAGCACT[A/G]TATCTAGGTACAAAGAACCTTGAAA
Phenotype
MIM: 300583
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VGLL1 PubMed Links

Gene Details

Gene
VGLL1
Gene Name
vestigial like family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016267.3 Intron NP_057351.1

View Full Product Details