Product Details

SNP ID
rs6040540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:1204115 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCAGATGGACATGCGTACACCCA[A/G]TGTCTTAGCCTGGGTCCCCTAGAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C20orf202 PubMed Links

Gene Details

Gene
C20orf202
Gene Name
chromosome 20 open reading frame 202
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009612.2 Intron NP_001009612.1

View Full Product Details