Product Details

SNP ID
rs9578604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:23472041 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTTTAAAAAGACGCGAAGCTATG[A/T]TCTTTTTATGAATATAGGTGTAAAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00327 PubMed Links

Gene Details

Gene
LINC00327
Gene Name
long intergenic non-protein coding RNA 327
There are no transcripts associated with this gene.

View Full Product Details