Product Details

SNP ID
rs10448313
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135346448 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGCCTGCATTCAGGCCAGGGGCC[A/G]TGTTTGCTCCCCCATTACAAGTTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf62 PubMed Links

Gene Details

Gene
C9orf62
Gene Name
chromosome 9 open reading frame 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173520.2 1810 UTR 3 NP_775791.1

View Full Product Details