Product Details
- SNP ID
-
rs10280920
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:83366854 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAATCTTTTACAGTAGTTAGAAT[C/T]CTAACTATAACTTCATCTGATTTGA
- Phenotype
-
MIM: 608166
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SEMA3E
PubMed Links
Gene Details
- Gene
- SEMA3E
- Gene Name
- semaphorin 3E
View Full Product Details