Product Details

SNP ID
rs9638807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:23618363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCCAGCTCCCCAACCCCCCATT[C/T]TGGGTCTTCCCATTAGCACAAAATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC126 PubMed Links

Gene Details

Gene
CCDC126
Gene Name
coiled-coil domain containing 126
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138771.3 Intron NP_620126.2
XM_017012775.1 Intron XP_016868264.1
XM_017012776.1 Intron XP_016868265.1

View Full Product Details