Product Details

SNP ID
rs7541117
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:166061514 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAACCAAATTTCAAATTCTTAGG[C/T]AGATGGTAAAAAAAACTTGTTCCAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM78B PubMed Links

Gene Details

Gene
FAM78B
Gene Name
family with sequence similarity 78 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017961.4 Intron NP_001017961.1
NM_001320302.1 Intron NP_001307231.1
XM_017000417.1 Intron XP_016855906.1
XM_017000418.1 Intron XP_016855907.1
Gene
LOC105371586
Gene Name
uncharacterized LOC105371586
There are no transcripts associated with this gene.

View Full Product Details