Product Details

SNP ID
rs6131924
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:17230569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTTCCCTTGAGACACAATTTTA[A/G]AGCTCTGCTATTTTCCCTTTTCTTT
Phenotype
MIM: 162151
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCSK2 PubMed Links

Gene Details

Gene
PCSK2
Gene Name
proprotein convertase subtilisin/kexin type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201528.1 Intron NP_001188457.1
NM_001201529.2 Intron NP_001188458.1
NM_002594.4 Intron NP_002585.2

View Full Product Details