Product Details

SNP ID
rs5967208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:100890521 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTAGGAACTCCAGCAAGTCCTTT[A/G]CATAAAAAATCCACATGAAATAGTT
Phenotype
MIM: 300684
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HNRNPA1P27 PubMed Links

Gene Details

Gene
HNRNPA1P27
Gene Name
heterogeneous nuclear ribonucleoprotein A1 pseudogene 27
There are no transcripts associated with this gene.

Gene
XKRX
Gene Name
XK related, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_212559.2 Intron NP_997724.2
XM_005262130.2 Intron XP_005262187.1
XM_011530954.2 Intron XP_011529256.1
XM_011530955.1 Intron XP_011529257.1
XM_017029517.1 Intron XP_016885006.1

View Full Product Details