Product Details
- SNP ID
-
rs10145029
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:34575559 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAAGTATAATAAAGAGGAAACTTT[C/T]ATGTAGGGTTTCCCACATGAAATAT
- Phenotype
-
MIM: 606098
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SNX6
PubMed Links
Gene Details
- Gene
- SNX6
- Gene Name
- sorting nexin 6
View Full Product Details