Product Details

SNP ID
hCV29527617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:176860866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGAGAAGGTTGTAGAGAGAAGA[A/T]GGGACTCACTACTATTATTTAATGA
Phenotype
MIM: 600904
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ASTN1 PubMed Links

Gene Details

Gene
ASTN1
Gene Name
astrotactin 1
There are no transcripts associated with this gene.

View Full Product Details